missing translation for 'onlineSavingsMsg'
Learn More

(Benzylthio)acetone, 98%, Thermo Scientific Chemicals

$195.95 - $195.95

Chemical Identifiers

CAS 10230-69-0
Molecular Formula C10H12OS
Molecular Weight (g/mol) 180.265
MDL Number MFCD00026241
InChI Key OIEDQMIEPJIRFT-UHFFFAOYSA-N
Synonym 1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
PubChem CID 82472
IUPAC Name 1-benzylsulfanylpropan-2-one
SMILES CC(=O)CSCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0381414
View Documents
Thermo Scientific Chemicals
L0381414
25 g
Each for $195.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10230-69-0
180.265
OIEDQMIEPJIRFT-UHFFFAOYSA-N
82472
CC(=O)CSCC1=CC=CC=C1
C10H12OS
MFCD00026241
1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
1-benzylsulfanylpropan-2-one

Specifications

10230-69-0
155°C to 156°C (17 mmHg)
C10H12OS
MFCD00026241
1866268
OIEDQMIEPJIRFT-UHFFFAOYSA-N
1-benzylsulfanylpropan-2-one
82472
98%
1.09
>100°C (212°F)
1.558
25 g
1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
CC(=O)CSCC1=CC=CC=C1
180.265
180.27
(Benzylthio)acetone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-552-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only