missing translation for 'onlineSavingsMsg'
Learn More

(Benzylthio)acetone, 98%, Thermo Scientific Chemicals

Catalog No. AAL0381414
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0381414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0381414 Supplier Thermo Scientific Chemicals Supplier No. L0381414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10230-69-0 | Molecular Formula: C10H12OS | Molecular Weight: 180.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10230-69-0
Molecular Formula C10H12OS
Molecular Weight (g/mol) 180.265
MDL Number MFCD00026241
InChI Key OIEDQMIEPJIRFT-UHFFFAOYSA-N
Synonym 1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
PubChem CID 82472
IUPAC Name 1-benzylsulfanylpropan-2-one
SMILES CC(=O)CSCC1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 155°C to 156°C (17 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.558
Quantity 25 g
Beilstein 1866268
Formula Weight 180.27
Percent Purity 98%
Chemical Name or Material (Benzylthio)acetone
EINECSNumber 233-552-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only