missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylsulfonyl Chloride 97.0+%, TCI America™
Supplier: TCI America B044125G
Specifications
| Benzylsulfonyl Chloride | |
| 92°C | |
| 120°C | |
| MFCD00007455 | |
| 3261 | |
| OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
| phenylmethanesulfonyl chloride | |
| 74740 | |
| ≥97.0% (T) |
| 1939-99-7 | |
| White-Yellow | |
| C7H7ClO2S | |
| 25 g | |
| alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
| C1=CC=C(C=C1)CS(=O)(=O)Cl | |
| 190.641 | |
| 190.64 | |
| Crystalline Powder |
Chemical Identifiers
| 1939-99-7 | |
| 190.641 | |
| OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
| 74740 | |
| C1=CC=C(C=C1)CS(=O)(=O)Cl |
| C7H7ClO2S | |
| MFCD00007455 | |
| alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
| phenylmethanesulfonyl chloride |
Safety and Handling
EINECSNumber : (3)-3482
TSCA : Yes