missing translation for 'onlineSavingsMsg'
Learn More

Benzylsulfonyl Chloride 97.0+%, TCI America™

Catalog No. p-7118446
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044125G 25 g
B0441100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B044125G Supplier TCI America Supplier No. B044125G
Only null left

Chemical Identifiers

CAS 1939-99-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00007455
InChI Key OAHKWDDSKCRNFE-UHFFFAOYSA-N
Synonym alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
PubChem CID 74740
IUPAC Name phenylmethanesulfonyl chloride
SMILES C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 3261
Formula Weight 190.64
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylsulfonyl Chloride
EINECSNumber (3)-3482
TSCA Yes