missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetyl chloride, 95%

Catalog No. AAL1433714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1433714 25 g
AAL1433706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1433714 Supplier Thermo Scientific Chemicals Supplier No. L1433714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19810-31-2 | C9H9ClO2 | 184.619 g/mol

Benzyloxyacetyl chloride is a reagent used to construct substituted 2-azetidinones for further elaboration into annulated β-lactams. It is a commonly employed reagent for asymmetric synthesis of β-lactams. It is also used in preparation of (S)-3-(methylamino)-3-((R)-pyrrolidin-3-yl)propanenitrile, key intermediate in the preparation of fluoroquinolone antibiotic for respiratory tract infections, non-racemic helicene and β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyloxyacetyl chloride is a reagent used to construct substituted 2-azetidinones for further elaboration into annulated β-lactams. It is a commonly employed reagent for asymmetric synthesis of β-lactams. It is also used in preparation of (S)-3-(methylamino)-3-((R)-pyrrolidin-3-yl)propanenitrile, key intermediate in the preparation of fluoroquinolone antibiotic for respiratory tract infections, non-racemic helicene and β-lactams.

Solubility
Reacts with water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas.

Chemical Identifiers

CAS 19810-31-2
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00010768
InChI Key QISAUDWTBBNJIR-UHFFFAOYSA-N
Synonym benzyloxyacetyl chloride, 2-benzyloxy acetyl chloride, benzyloxy acetyl chloride, benzyloxyacetylchloride, benzyloxy-acetyl chloride, benzyloxyacetic acid chloride, acetyl chloride, phenylmethoxy, acetyl chloride, benzyloxy, alpha-benzyloxy acetyl chloride, bnoch2cocl
PubChem CID 177085
IUPAC Name 2-phenylmethoxyacetyl chloride
SMILES C1=CC=C(C=C1)COCC(=O)Cl

Specifications

Density 1.189
Boiling Point 84°C to 87°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.523
Linear Formula C6H5CH2OCH2COCl
Quantity 25 g
UN Number UN3265
Beilstein 1947363
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 184.62
Percent Purity 95%
Chemical Name or Material Benzyloxyacetyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only