missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetic Acid 97.0+%, TCI America™

Catalog No. B28761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28761G 1 g
B28765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28761G Supplier TCI America Supplier No. B28761G

Chemical Identifiers

CAS 30379-55-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00274211
InChI Key GRZHHTYDZVRPIC-UHFFFAOYSA-N
Synonym benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
PubChem CID 290301
IUPAC Name 2-(benzyloxy)acetic acid
SMILES OC(=O)COCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzyloxyacetic Acid