missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetic acid, 95%

Catalog No. AAH5458106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5458106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5458106 Supplier Thermo Scientific Chemicals Supplier No. H5458106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30379-55-6 | C9H10O3 | 166.18 g/mol

It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterification, tetra aqua(1,10-phenanthroline-[κ]2N,N?)magnesium(II) bis[(2,4-dichlorophenyl)acetate], as ligand in the preparation of diaquabis(benzyloxyacetato)copper(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterific

Chemical Identifiers

CAS 30379-55-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00274211
InChI Key GRZHHTYDZVRPIC-UHFFFAOYSA-N
Synonym benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
PubChem CID 290301
IUPAC Name 2-phenylmethoxyacetic acid
SMILES OC(=O)COCC1=CC=CC=C1

Specifications

Density 1.162
Boiling Point 137°C to 139°C
Flash Point 113°C (235°F)
Refractive Index 1.526
Linear Formula C6H5CH2OCH2COOH
Quantity 5 g
Formula Weight 166.17
Percent Purity 95%
Chemical Name or Material Benzyloxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 608-470-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only