Learn More
Benzyloxyacetaldehyde diethyl acetal, 98%
CAS: 42783-78-8 | C13H20O3 | 224.3 g/mol
Supplier: Thermo Scientific Chemicals A1447418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Benzyloxyacetaldehyde diethyl acetal | |
| 0.987 | |
| 79°C (174°F) | |
| 1.479 | |
| 50 g | |
| 2,2-diethoxyethoxy methyl benzene, benzyloxyacetaldehyde diethyl acetal, benzene, 2,2-diethoxyethoxy methyl, acmc-20alp5, 1,1-diethoxy-2-benzyloxy ethane, 2,2-diethoxy-ethoxymethyl-benzene, 2,2-diethoxy-1-phenylmethoxy ethane, 2-benzyloxy acetaldehyde diethyl acetal | |
| CCOC(COCC1=CC=CC=C1)OCC | |
| 224.3 | |
| 224.3 |
| 42783-78-8 | |
| 99°C to 100°C (0.6 mmHg) | |
| C13H20O3 | |
| MFCD00051786 | |
| 3278795 | |
| VTYTZCJKJNWMGA-UHFFFAOYSA-N | |
| 2,2-diethoxyethoxymethylbenzene | |
| 4321487 | |
| 98% |
Chemical Identifiers
| 42783-78-8 | |
| 224.3 | |
| VTYTZCJKJNWMGA-UHFFFAOYSA-N | |
| 4321487 | |
| CCOC(COCC1=CC=CC=C1)OCC |
| C13H20O3 | |
| MFCD00051786 | |
| 2,2-diethoxyethoxy methyl benzene, benzyloxyacetaldehyde diethyl acetal, benzene, 2,2-diethoxyethoxy methyl, acmc-20alp5, 1,1-diethoxy-2-benzyloxy ethane, 2,2-diethoxy-ethoxymethyl-benzene, 2,2-diethoxy-1-phenylmethoxy ethane, 2-benzyloxy acetaldehyde diethyl acetal | |
| 2,2-diethoxyethoxymethylbenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only