missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde diethyl acetal, 98%

Catalog No. aaa1447409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1447409 10 g
AAA1447418 50 g
AAA1447430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1447409 Supplier Thermo Scientific Chemicals Supplier No. A1447409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42783-78-8 | C13H20O3 | 224.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42783-78-8
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD00051786
InChI Key VTYTZCJKJNWMGA-UHFFFAOYSA-N
Synonym 2,2-diethoxyethoxy methyl benzene, benzyloxyacetaldehyde diethyl acetal, benzene, 2,2-diethoxyethoxy methyl, acmc-20alp5, 1,1-diethoxy-2-benzyloxy ethane, 2,2-diethoxy-ethoxymethyl-benzene, 2,2-diethoxy-1-phenylmethoxy ethane, 2-benzyloxy acetaldehyde diethyl acetal
PubChem CID 4321487
IUPAC Name 2,2-diethoxyethoxymethylbenzene
SMILES CCOC(COCC1=CC=CC=C1)OCC

Specifications

Density 0.987
Boiling Point 99°C to 100°C (0.6 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.479
Quantity 10 g
Beilstein 3278795
Formula Weight 224.3
Percent Purity 98%
Chemical Name or Material Benzyloxyacetaldehyde diethyl acetal
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only