missing translation for 'onlineSavingsMsg'
Learn More

Benzylidenemalononitrile, 98+%

Catalog No. AAL0242614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0242614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0242614 Supplier Thermo Scientific Chemicals Supplier No. L0242614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2700-22-3 | C10H6N2 | 154.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2700-22-3
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00001855
InChI Key WAVNYPVYNSIHNC-UHFFFAOYSA-N
Synonym benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene
PubChem CID 17608
IUPAC Name 2-benzylidenepropanedinitrile
SMILES C1=CC=C(C=C1)C=C(C#N)C#N

Specifications

Melting Point 83°C to 85°C
Boiling Point 120°C to 125°C (0.6 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 1907519
Formula Weight 154.17
Percent Purity ≥98%
Chemical Name or Material Benzylidenemalononitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 220-283-6
RTECSNumber OO3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only