accessibility menu, dialog, popup

UserName

NN

CAS RN 2700-22-3

IUPAC Name: 2-(phenylmethylidene)propanedinitrile
Synonyms: malononitrile, benzal
Molecular Weight (g/mol): 154.17
Molecular Formula: C10H6N2
InChi Key: WAVNYPVYNSIHNC-UHFFFAOYSA-N
SMILES: N#CC(=CC1=CC=CC=C1)C#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)