missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine dihydrochloride, 97%

Catalog No. p-66025
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105960050 5 g Glass bottle
AC105960250 25 g Glass bottle
AC105961000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105960050 Supplier Thermo Scientific Chemicals Supplier No. 105960050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20570-96-1 | C7H10N2·2HCl | 195.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20570-96-1
Molecular Formula C7H10N2·2HCl
Molecular Weight (g/mol) 195.09
MDL Number MFCD00012921
InChI Key MSJHOJKVMMEMNX-UHFFFAOYSA-N
Synonym benzylhydrazine dihydrochloride, benzylhydrazine.2hcl, 1-benzylhydrazine dihydrochloride, benzyl-hydrazine dihydrochloride, hydrazine, phenylmethyl-, dihydrochloride, hydrazine, benzyl-, dihydrochloride, phenylmethyl hydrazine dihydrochloride, benzyl hydrazine di hcl, benzyl-hydrazine hydrochloride, benzylhydrazinedihydrochloride
PubChem CID 146540
IUPAC Name benzylhydrazine;dihydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 143°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2NHNH2·2HCl
Quantity 5 g
Beilstein 15,531
Formula Weight 195.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Benzylhydrazine dihydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 243-887-1
RTECSNumber MU8540000

RUO – Research Use Only