missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine dihydrochloride, 97%

Catalog No. p-7036598
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5491606 5 g
AAH5491614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5491606 Supplier Thermo Scientific Chemicals Supplier No. H5491606
Only null left

CAS: 20570-96-1 | C7H12Cl2N2 | 195.087 g/mol

Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20570-96-1
Molecular Formula C7H12Cl2N2
Molecular Weight (g/mol) 195.087
MDL Number MFCD00012921
InChI Key MSJHOJKVMMEMNX-UHFFFAOYSA-N
Synonym benzylhydrazine dihydrochloride, benzylhydrazine.2hcl, 1-benzylhydrazine dihydrochloride, benzyl-hydrazine dihydrochloride, hydrazine, phenylmethyl-, dihydrochloride, hydrazine, benzyl-, dihydrochloride, phenylmethyl hydrazine dihydrochloride, benzyl hydrazine di hcl, benzyl-hydrazine hydrochloride, benzylhydrazinedihydrochloride
PubChem CID 146540
IUPAC Name benzylhydrazine;dihydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl.Cl

Specifications

Melting Point 143°C to 145°C
Linear Formula C6H5CH2NHNH2·2HCl
Quantity 5 g
UN Number UN2811
Beilstein 3688990
Solubility Information Insoluble in water.
Formula Weight 195.09
Percent Purity 97%
Chemical Name or Material Benzylhydrazine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 243-887-1
RTECSNumber MU8540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only