missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyldimethyltetradecylammonium Chloride Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 100 g |
| UN Number | 2923 |
| Formula Weight | 368.05 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzyldimethyltetradecylammonium Chloride Hydrate |
Safety and Handling
| EINECSNumber | (3)-2694 |
| RTECSNumber | BO7150000 |
| TSCA | Yes |