missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride hydrate, 97%

Catalog No. p-66010
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402070050 5 g Glass bottle
AC402075000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402070050 Supplier Thermo Scientific Chemicals Supplier No. 402070050
Only null left

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color Cream-White to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on anhydrous substance) (Argentometry)
Packaging Glass bottle
Quantity 5 g
Beilstein 12, III, 2212
Merck Index 15, 2016
Solubility Information Solubility in water: 8.5g/L. Other solubilities: soluble in alcohol, acetone, esters, ethyla acetat, e, propylene glycol, sorbitol solutions, glycerol,, ether
Formula Weight 396.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate, 97 - 101%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 204-526-3
RTECSNumber BO6822450
TSCA TSCA

RUO – Research Use Only