accessibility menu, dialog, popup

UserName

CAS RN 122-18-9

IUPAC Name: benzyl(hexadecyl)dimethylazanium chloride
Synonyms: cetalkonium chloride|alkyl* dimethyl benzyl ammonium chloride *(100% C16)
Molecular Weight (g/mol): 396.1
Molecular Formula: C25H46ClN
InChi Key: SXPWTBGAZSPLHA-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)