missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride, 95%

Catalog No. p-7049971
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3237718 50 g
AA3237730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3237718 Supplier Thermo Scientific Chemicals Supplier No. 03237718
Only null left

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol

Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 54°C to 58°C
Color White
Linear Formula CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
Quantity 50 g
UN Number UN3261
Beilstein 3802341
Sensitivity Hygroscopic
Merck Index 14,2017
Solubility Information Soluble in water (8.5g/L),ethanol,acetone,ethyl acetate,propylene glycol,sorbitol solutions,ether,glycerol,and carbon tetrachloride.
Formula Weight 396.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-526-3
RTECSNumber BO6822450
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only