missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyl-n-tetradecylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Phase transfer catalyst

$88.10 - $88.10

Chemical Identifiers

CAS 139-08-2
Molecular Formula C23H42ClN
Molecular Weight (g/mol) 368.05
MDL Number MFCD00011771
InChI Key OCBHHZMJRVXXQK-UHFFFAOYSA-M
Synonym miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
PubChem CID 8755
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6346514
View Documents
Thermo Scientific Chemicals
J6346514
25 g
Each for $88.10
Only null left
 
Description

Description

Phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

139-08-2
368.05
OCBHHZMJRVXXQK-UHFFFAOYSA-M
8755
C23H42ClN
MFCD00011771
miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
[Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

139-08-2
C23H42ClN
MFCD00011771
UN3261
14,1059
OCBHHZMJRVXXQK-UHFFFAOYSA-M
benzyl-dimethyl-tetradecylazanium;chloride
8755
98%
Benzyldimethyl-n-tetradecylammonium chloride hydrate
White
CH3(CH2)13N(CH3)2(CH2C6H5)Cl
25 g
4062599
miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
[Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
368.05
368.05 (Anhydrous)
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 205-352-0

RTECSNumber : BO7150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only