missing translation for 'onlineSavingsMsg'
Learn More

Benzylcetyldimethylammonium Chloride Hydrate 97.0+%, TCI America™

Catalog No. B0237500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0237500G 500 g
B023725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0237500G Supplier TCI America Supplier No. B0237500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-18-9
Molecular Formula C25H46ClN
Molecular Weight (g/mol) 396.10
MDL Number MFCD00149967
InChI Key SXPWTBGAZSPLHA-UHFFFAOYSA-M
Synonym cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
PubChem CID 31202
IUPAC Name benzyl(hexadecyl)dimethylazanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 65°C
Color White
Quantity 500 g
Formula Weight 396.10
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzylcetyldimethylammonium Chloride Hydrate
EINECSNumber (3)-2694
RTECSNumber BO6822450
TSCA Yes