missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylbis(triphenylphosphine)palladium(II) Chloride 96.0+%, TCI America™
$63.38 - $378.40
Chemical Identifiers
| CAS | 22784-59-4 |
|---|---|
| Molecular Formula | C43H37ClP2Pd |
| Molecular Weight (g/mol) | 757.59 |
| MDL Number | MFCD00010395 |
| InChI Key | GYAXNTWMUGUEJY-UHFFFAOYSA-M |
| Synonym | Benzyl(chloro)bis(triphenylphosphine)palladium(II) |
| IUPAC Name | benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane) |
| SMILES | Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2029100MG
|
TCI America
B2029100MG |
100 mg |
Each for $63.38
|
|
|||||
|
B20291G
|
TCI America
B20291G |
1 g |
Each for $378.40
|
|
|||||
Chemical Identifiers
| 22784-59-4 | |
| 757.59 | |
| GYAXNTWMUGUEJY-UHFFFAOYSA-M | |
| benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane) |
| C43H37ClP2Pd | |
| MFCD00010395 | |
| Benzyl(chloro)bis(triphenylphosphine)palladium(II) | |
| Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 22784-59-4 | |
| MFCD00010395 | |
| Benzyl(chloro)bis(triphenylphosphine)palladium(II) | |
| Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 757.59 | |
| ≥96.0% (T) | |
| Benzylbis(triphenylphosphine)palladium(II) Chloride |
| C43H37ClP2Pd | |
| 100 mg | |
| GYAXNTWMUGUEJY-UHFFFAOYSA-M | |
| benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane) | |
| 757.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer