missing translation for 'onlineSavingsMsg'
Learn More

Benzylbis(triphenylphosphine)palladium(II) Chloride 96.0+%, TCI America™

Catalog No. B2029100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2029100MG 100 mg
B20291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2029100MG Supplier TCI America Supplier No. B2029100MG
Only null left

Chemical Identifiers

CAS 22784-59-4
Molecular Formula C43H37ClP2Pd
Molecular Weight (g/mol) 757.59
MDL Number MFCD00010395
InChI Key GYAXNTWMUGUEJY-UHFFFAOYSA-M
Synonym Benzyl(chloro)bis(triphenylphosphine)palladium(II)
IUPAC Name benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane)
SMILES Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 757.59
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylbis(triphenylphosphine)palladium(II) Chloride
TSCA No
Recommended Storage Freezer