missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine Hydroiodide (Low water content) 98.0+%, TCI America™

Catalog No. p-7123622
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B45661G 1 g
B45665G 5 g
2 options

Catalog No. B45661G

Supplier: TCI America B45661G

Only null left

Chemical Identifiers

CAS 45579-91-7
Molecular Formula C7H10IN
Molecular Weight (g/mol) 235.068
InChI Key PPCHYMCMRUGLHR-UHFFFAOYSA-N
Synonym Benzylammonium Iodide
PubChem CID 67825834
IUPAC Name phenylmethanamine;hydroiodide
SMILES C1=CC=C(C=C1)CN.I

Specifications

Melting Point 178°C
Quantity 1 g
Formula Weight 235.07
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzylamine Hydroiodide (Low water content)