Learn More
Benzylamine, 99.5+%, AcroSeal™
CAS: 100-46-9 | C7H9N | 107.15 g/mol
$173.97 - $173.97
Chemical Identifiers
| CAS | 100-46-9 |
|---|---|
| Molecular Formula | C7H9N |
| Molecular Weight (g/mol) | 107.15 |
| MDL Number | MFCD00008106 |
| InChI Key | WGQKYBSKWIADBV-UHFFFAOYSA-N |
| Synonym | benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine |
| PubChem CID | 7504 |
| ChEBI | CHEBI:40538 |
| IUPAC Name | phenylmethanamine |
| SMILES | C1=CC=C(C=C1)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC427231000
|
Thermo Scientific Chemicals
427231000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $173.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 100-46-9 | |
| 107.15 | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| 7504 | |
| phenylmethanamine |
| C7H9N | |
| MFCD00008106 | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| CHEBI:40538 | |
| C1=CC=C(C=C1)CN |
Specifications
| 100-46-9 | |
| 100.0 | |
| Colorless to Yellow | |
| 182.0°C to 185.0°C | |
| Authentic | |
| AcroSeal™ Glass Bottle | |
| 1.5424 to 1.5444 | |
| MFCD00008106 | |
| 0.98 | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| phenylmethanamine | |
| 7504 | |
| CHEBI:40538 | |
| 99.5+% | |
| Benzylamine |
| 99.5 | |
| -30.0°C | |
| 0.9800g/mL | |
| 72°C | |
| 99.5% min. (GC) | |
| C7H9N | |
| C6H5CH2NH2 | |
| 100 mL | |
| 15, 1128 | |
| Solubility in water: soluble in water. Other solubilities: >50g/100mL methanol,>50g/100mL ethanol,>50g/100mL 2-propanol,miscible with alcohol,ether,acetone,benzene | |
| C1=CC=C(C=C1)CN | |
| 107.15 | |
| 1.82 mPa.s (20°C) | |
| 107.15 | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 202-854-1
RUO – Research Use Only