Learn More
Benzylamine, 98+%
CAS: 100-46-9 | C7H9N | 107.156 g/mol
Supplier: Thermo Scientific Chemicals A109970B
Description
Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.
Solubility
Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
| Benzylamine | |
| -43°C | |
| 184°C to 185°C | |
| C7H9N | |
| MFCD00008106 | |
| UN2735 | |
| Air Sensitive | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| phenylmethanamine | |
| 7504 | |
| 107.16 |
| 100-46-9 | |
| 0.982 | |
| 72°C (162°F) | |
| 1.5435 | |
| 1000 g | |
| 741984 | |
| 14,1125 | |
| Miscible with water,ethanol,diethyl ether,acetone,benzene and chloroform. | |
| C1=CC=C(C=C1)CN | |
| 107.156 | |
| CHEBI:40538 | |
| ≥98% |
Chemical Identifiers
| 100-46-9 | |
| 107.156 | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| 7504 | |
| phenylmethanamine |
| C7H9N | |
| MFCD00008106 | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| CHEBI:40538 | |
| C1=CC=C(C=C1)CN |
Safety and Handling
GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P374-P501c
H227-H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 202-854-1
RTECSNumber : DP1488500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only