missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Thiocyanate 99.0+%, TCI America™

Catalog No. T019825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T019825G 25 g
T0198500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T019825G Supplier TCI America Supplier No. T019825G
Only null left

Chemical Identifiers

CAS 3012-37-1
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00001832
InChI Key ABNDFSOIUFLJAH-UHFFFAOYSA-N
Synonym tropeolin, thiocyanic acid, phenylmethyl ester, solvat 14, benzylthiocyanate, benzylrhodonid, thiocyanic acid, benzyl ester, alpha-thiocyanatotoluene, benzyl-thiocyanate, thiocyanic acid benzyl ester, phenylmethyl thiocyanate
PubChem CID 18170
ChEBI CHEBI:16017
IUPAC Name (benzylsulfanyl)carbonitrile
SMILES N#CSCC1=CC=CC=C1

Specifications

Melting Point 42°C
Color White
Boiling Point 256°C
Quantity 25 g
UN Number 1759
Formula Weight 149.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Thiocyanate
RTECSNumber XK8155000
TSCA Yes