missing translation for 'onlineSavingsMsg'
Learn More

Benzyl thioacetimidate hydrochloride, 96%

Catalog No. p-7034179
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5184403 1 g
AAH5184406 5 g
AAH5184414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5184403 Supplier Thermo Scientific Chemicals Supplier No. H5184403

CAS: 32894-07-8 | C9H12ClNS | 201.712 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32894-07-8
Molecular Formula C9H12ClNS
Molecular Weight (g/mol) 201.712
MDL Number MFCD05865210
InChI Key OVQWHRFBVPDPCD-UHFFFAOYSA-N
Synonym s-benzylthioacetimidate, hydrochloride, benzyl thioacetimidate hydrochloride, benzyl thioacetimidate hcl, benzyl ethanimidothioate hydrochloride, 1-benzylsulfanyl ethanimine hydrochloride, acmc-20ajr8, s-benzylthioacetimidate hydrochloride, thioacetimidic acid benzyl ester; hydrochloride, benzyl ethanimidothioate-hydrogen chloride 1/1
PubChem CID 12626231
IUPAC Name benzyl ethanimidothioate;hydrochloride
SMILES CC(=N)SCC1=CC=CC=C1.Cl

Specifications

Quantity 1 g
Formula Weight 201.71
Percent Purity 96%
Chemical Name or Material Benzyl thioacetimidate hydrochloride
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only