missing translation for 'onlineSavingsMsg'
Learn More

Benzyl tert-butyl malonate, 95%

Catalog No. AAL1553306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1553306 5 g
AAL1553314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1553306 Supplier Thermo Scientific Chemicals Supplier No. L1553306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 72594-86-6 | C14H18O4 | 250.294 g/mol

Benzyl tert-butyl malonate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl tert-butyl malonate is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 72594-86-6
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD01075175
InChI Key XKXXXODAXXAFNP-UHFFFAOYSA-N
Synonym benzyl tert-butyl malonate, benzyltert-butylmalonate, tert-butyl benzyl malonate, 1-benzyl 3-tert-butyl propanedioate, tert-butyl 2-benzyloxycarbonyl acetate, propanedioic acid, 1,1-dimethylethyl phenylmethyl ester, tert-butyl phenylmethyl propane-1,3-dioate, pubchem3938, t-butyl benzyl malonate, ksc493q0n
PubChem CID 2736712
IUPAC Name 1-O-benzyl 3-O-tert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Refractive Index 1.486
Quantity 5 g
Beilstein 5434466
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 250.3
Percent Purity 95%
Chemical Name or Material Benzyl tert-butyl malonate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only