accessibility menu, dialog, popup

UserName

CAS RN 72594-86-6

IUPAC Name: 1-benzyl 3-tert-butyl propanedioate
Synonyms: 1-benzyl 3-tert-butyl propanedioate
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: XKXXXODAXXAFNP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results