missing translation for 'onlineSavingsMsg'
Learn More

Benzyl sulfide, 98%, Thermo Scientific™

$51.31 - $51.31

Chemical Identifiers

CAS 538-74-9
Molecular Formula C14H14S
Molecular Weight (g/mol) 214.33
MDL Number MFCD00004784
InChI Key LUFPJJNWMYZRQE-UHFFFAOYSA-N
Synonym dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
PubChem CID 10867
IUPAC Name benzylsulfanylmethylbenzene
SMILES C(SCC1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC335000050
SDS View Documents
Thermo Scientific Chemicals
335000050
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

538-74-9
214.33
LUFPJJNWMYZRQE-UHFFFAOYSA-N
10867
C(SCC1=CC=CC=C1)C1=CC=CC=C1
C14H14S
MFCD00004784
dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
benzylsulfanylmethylbenzene

Specifications

538-74-9
100.0
White
Authentic
Glass bottle
MFCD00004784
15, 1147
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether
C(SCC1=CC=CC=C1)C1=CC=CC=C1
214.33
214.3
Crystals
97.5
44°C to 49°C
>110°C
97.5% min. (GC)
C14H14S
5 g
dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
LUFPJJNWMYZRQE-UHFFFAOYSA-N
benzylsulfanylmethylbenzene
10867
98%
Benzyl sulfide, 98%
Videos
Safety and Handling

Safety and Handling

STENCH

EINECSNumber : 208-703-6

RTECSNumber : DC0512500

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only