missing translation for 'onlineSavingsMsg'
Learn More

Benzyl salicylate, 99%

CAS: 118-58-1 | C14H12O3 | 228.247 g/mol

Supplier:  Thermo Scientific Chemicals A161840E

 View more versions of this product

Catalog No. AAA161840E

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzyl salicylate
18°C to 20°C
190°C (14 mmHg)
C14H12O3
MFCD00020034
2115365
benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O
228.247
228.25
118-58-1
1.17
137°C (278°F)
1.5804
2500 g
14,1144
ZCTQGTTXIYCGGC-UHFFFAOYSA-N
benzyl 2-hydroxybenzoate
8363
99%

Chemical Identifiers

118-58-1
228.247
ZCTQGTTXIYCGGC-UHFFFAOYSA-N
8363
C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O
C14H12O3
MFCD00020034
benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
benzyl 2-hydroxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 204-262-9

RTECSNumber : VO1750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only