missing translation for 'onlineSavingsMsg'
Learn More

Benzyl salicylate, 99%

Catalog No. AAA1618436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1618436 500 g
AAA1618422 100 g
AAA161840E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1618436 Supplier Thermo Scientific Chemicals Supplier No. A1618436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-58-1 | C14H12O3 | 228.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-58-1
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00020034
InChI Key ZCTQGTTXIYCGGC-UHFFFAOYSA-N
Synonym benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
PubChem CID 8363
IUPAC Name benzyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O

Specifications

Melting Point 18°C to 20°C
Density 1.17
Boiling Point 190°C (14 mmHg)
Flash Point 137°C (278°F)
Refractive Index 1.5804
Quantity 500 g
Beilstein 2115365
Merck Index 14,1144
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl salicylate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-262-9
RTECSNumber VO1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only