missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals

Catalog No. AAH6291206
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6291206 5 g
AAH62912MD 250 mg
AAH6291203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6291206 Supplier Thermo Scientific Chemicals Supplier No. H6291206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113400-36-5
Molecular Formula C17H21NO5
Molecular Weight (g/mol) 319.357
MDL Number MFCD04115781
InChI Key TZNBTMCEMLXYEM-ZDUSSCGKSA-N
Synonym s-2-benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate, boc-pyr-obzl, boc-l-pyroglutamic acid benzyl ester, 2-benzyl 1-tert-butyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pyroglutamic acid benzyl ester, 2-benzyl 1-tert-butyl s-5-oxopyrrolidine-1,2-dicarboxylate, benzyl n-boc-l-pyroglutamate, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl 2-phenylmethyl ester, s, n-boc-l-pyroglutamic acid benzyl ester, benzyl n-tert-butoxycarbonyl-5-oxoprolinate
PubChem CID 8027482
IUPAC Name 2-O-benzyl 1-O-tert-butyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OCC2=CC=CC=C2

Specifications

Color White
Quantity 5 g
Formula Weight 319.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only