accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOO

CAS RN 113400-36-5

IUPAC Name: 2-benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 2-benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 319.36
Molecular Formula: C17H21NO5
InChi Key: TZNBTMCEMLXYEM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)