missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Propionate 99.0+%, TCI America™

Catalog No. p-7134641
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050125ML 25 mL
P0501500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050125ML Supplier TCI America Supplier No. P050125ML
Only null left

Chemical Identifiers

CAS 122-63-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00027003
InChI Key VHOMAPWVLKRQAZ-UHFFFAOYSA-N
Synonym benzyl propionate, propanoic acid, phenylmethyl ester, propionic acid, benzyl ester, phenylmethyl propanoate, propionic acid benzyl ester, phenylmethyl propionate, benzyl propionate natrual, benzyl n-propionate, fema no. 2150, propionic acid, benzyl ester 6ci,7ci,8ci
PubChem CID 31219
IUPAC Name benzyl propanoate
SMILES CCC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 222°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Propionate
EINECSNumber (3)-1020
TSCA Yes