CAS RN 122-63-4
CAS RN 122-63-4
IUPAC Name:
benzyl propanoate
Synonyms:
benzyl propionate
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
VHOMAPWVLKRQAZ-UHFFFAOYSA-N
SMILES:
CCC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Benzyl Propionate 99.0+%, TCI America™
CAS: 122-63-4 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.20 MDL Number: MFCD00027003 InChI Key: VHOMAPWVLKRQAZ-UHFFFAOYSA-N Synonym: benzyl propionate,propanoic acid, phenylmethyl ester,propionic acid, benzyl ester,phenylmethyl propanoate,propionic acid benzyl ester,phenylmethyl propionate,benzyl propionate natrual,benzyl n-propionate,fema no. 2150,propionic acid, benzyl ester 6ci,7ci,8ci PubChem CID: 31219 IUPAC Name: benzyl propanoate SMILES: CCC(=O)OCC1=CC=CC=C1