missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. p-7119935
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30805G 5 g
B308025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30805G Supplier TCI America Supplier No. B30805G
Only null left

Chemical Identifiers

CAS 3112-88-7
Molecular Formula C13H12O2S
Molecular Weight (g/mol) 232.297
MDL Number MFCD00025040
InChI Key FABCMLOTUSCWOR-UHFFFAOYSA-N
Synonym benzyl phenyl sulfone, benzylsulfonyl benzene, sulfone, benzyl phenyl, benzylphenylsulfone, phenyl benzyl sulfone, benzyl phenyl sulphone, benzene, phenylmethyl sulfonyl, phenylmethyl sulfonyl benzene, phenylmethyl sulphonyl benzene, benzenesulfonyl methyl benzene
PubChem CID 76561
IUPAC Name benzenesulfonylmethylbenzene
SMILES C1=CC=C(C=C1)CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 149°C
Color White
Quantity 5 g
Formula Weight 232.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Phenyl Sulfone
RTECSNumber WR2700000
TSCA No