missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Phenyl Ether 98.0+%, TCI America™

Catalog No. B0433500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0433500G 500 g
B043325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0433500G Supplier TCI America Supplier No. B0433500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 946-80-5
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00020660
InChI Key BOTNYLSAWDQNEX-UHFFFAOYSA-N
Synonym benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
PubChem CID 70352
IUPAC Name phenoxymethylbenzene
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 288°C
Quantity 500 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Phenyl Ether
TSCA No
Recommended Storage Refrigerator