missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Phenyl Carbonate 95.0+%, TCI America™

Catalog No. p-7123023
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35745G 5 g
B357425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35745G Supplier TCI America Supplier No. B35745G
Only null left

Chemical Identifiers

CAS 28170-07-2
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD07784341
InChI Key SNGLYCMNDNOLOF-UHFFFAOYSA-N
Synonym Carbonic Acid Benzyl Phenyl Ester
PubChem CID 11183772
IUPAC Name benzyl phenyl carbonate
SMILES C1=CC=C(C=C1)COC(=O)OC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 g
Formula Weight 228.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Phenyl Carbonate