accessibility menu, dialog, popup

UserName

CAS RN 28170-07-2

OOO

CAS RN 28170-07-2

IUPAC Name: benzyl phenyl carbonate
Synonyms: benzyl phenyl carbonate
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: SNGLYCMNDNOLOF-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results