missing translation for 'onlineSavingsMsg'
Learn More

Benzyl n-butyl phthalate, 98%

Catalog No. p-7027764
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2476922 100 g
AAB2476936 500 g
AAB247690E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2476922 Supplier Thermo Scientific Chemicals Supplier No. B2476922
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 85-68-7 | C19H20O4 | 312.365 g/mol

Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, alkalis and strong bases.

Chemical Identifiers

CAS 85-68-7
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.365
MDL Number MFCD00009440
InChI Key IRIAEXORFWYRCZ-UHFFFAOYSA-N
Synonym benzyl butyl phthalate, butyl benzyl phthalate, sicol, palatinol bb, unimoll bb, santicizer 160, butylbenzyl phthalate, n-butyl benzyl phthalate, sicol 160, benzyl n-butyl phthalate
PubChem CID 2347
ChEBI CHEBI:34595
IUPAC Name 2-O-benzyl 1-O-butyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCC2=CC=CC=C2

Specifications

Density 1.1
Boiling Point 370°C
Flash Point >110°C (230°F)
Refractive Index 1.54
Quantity 100 g
UN Number UN3082
Solubility Information Slightly miscible with water.
Formula Weight 312.37
Percent Purity 98%
Chemical Name or Material Benzyl n-butyl phthalate
Hazard Category H319-H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-622-7
TSCA Yes

RUO – Research Use Only