missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Methyl Sulfide 98.0+%, TCI America™
Supplier: TCI America B099225ML
Specifications
| Benzyl Methyl Sulfide | |
| Yellow | |
| C8H10S | |
| 25 mL | |
| OFQPKKGMNWASPN-UHFFFAOYSA-N | |
| methylsulfanylmethylbenzene | |
| 13016 | |
| ≥98.0% (GC) |
| 766-92-7 | |
| 200°C | |
| MFCD00008563 | |
| benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51 | |
| CSCC1=CC=CC=C1 | |
| 138.228 | |
| 138.23 | |
| Liquid |
Chemical Identifiers
| 766-92-7 | |
| 138.228 | |
| OFQPKKGMNWASPN-UHFFFAOYSA-N | |
| 13016 | |
| CSCC1=CC=CC=C1 |
| C8H10S | |
| MFCD00008563 | |
| benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51 | |
| methylsulfanylmethylbenzene |
Safety and Handling
TSCA : Yes