missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Methyl Sulfide 98.0+%, TCI America™

Catalog No. B09925ML
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
B09925ML 5 mL
B099225ML 25 mL
2 options

Catalog No. B09925ML

Supplier: TCI America B09925ML

Only null left

Chemical Identifiers

CAS 766-92-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00008563
InChI Key OFQPKKGMNWASPN-UHFFFAOYSA-N
Synonym benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51
PubChem CID 13016
IUPAC Name methylsulfanylmethylbenzene
SMILES CSCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 200°C
Quantity 5 mL
Formula Weight 138.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Methyl Sulfide