missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Methyl Ether 97.0+%, TCI America™

Catalog No. B042425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B042425ML 25 mL
B0424500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B042425ML Supplier TCI America Supplier No. B042425ML

Chemical Identifiers

CAS 538-86-3
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00025901
InChI Key GQKZBCPTCWJTAS-UHFFFAOYSA-N
Synonym benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
PubChem CID 10869
IUPAC Name methoxymethylbenzene
SMILES COCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 174°C
Quantity 25 mL
UN Number 3271
Formula Weight 122.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Methyl Ether