missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methyl ether, 97%

Catalog No. p-7027043
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349009 10 g
AAB2349018 50 g
AAB2349030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2349009 Supplier Thermo Scientific Chemicals Supplier No. B2349009

CAS: 538-86-3 | C8H10O | 122.167 g/mol

Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 538-86-3
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00025901
InChI Key GQKZBCPTCWJTAS-UHFFFAOYSA-N
Synonym benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
PubChem CID 10869
IUPAC Name methoxymethylbenzene
SMILES COCC1=CC=CC=C1

Specifications

Melting Point -53°C
Density 0.94
Boiling Point 173°C to 175°C
Flash Point 56°C (133°F)
Refractive Index 1.5015
Quantity 10 g
UN Number UN3271
Merck Index 14,1140
Solubility Information Insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material Benzyl methyl ether
Show More Show Less
Hazard Category H226-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 208-705-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only