missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Mercaptan 96.0+%, TCI America™

Catalog No. T028725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T028725G 25 g
T0287100G 100 g
T0287500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T028725G Supplier TCI America Supplier No. T028725G

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Melting Point -30°C
Color Yellow
Boiling Point 195°C
Quantity 25 g
UN Number 2810
Formula Weight 124.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Mercaptan
EINECSNumber (3)-1105
RTECSNumber XT8650000
TSCA Yes