accessibility menu, dialog, popup

UserName

HS

CAS RN 100-53-8

IUPAC Name: phenylmethanethiol
Synonyms: benzyl mercaptan
Molecular Weight (g/mol): 124.2
Molecular Formula: C7H8S
InChi Key: UENWRTRMUIOCKN-UHFFFAOYSA-N
SMILES: SCC1=CC=CC=C1
Physical Form 
  • (1)
  • (7)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (5)
  • (1)
  • (7)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)