missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, 99%

Catalog No. AAB2008114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2008114 25 g
AAB2008122 100 g
AAB2008136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2008114 Supplier Thermo Scientific Chemicals Supplier No. B2008114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-53-8 | C7H8S | 124.20 g/mol

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.053
Boiling Point 194°C to 195°C
Flash Point 70°C (158°F)
Odor Stench
Assay Percent Range 0.99
Refractive Index 1.576
Quantity 25 g
UN Number UN2810
Beilstein 605864
Sensitivity Air Sensitive
Merck Index 14,9322
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less
Hazard Category H227-H302-H319-H330
Hazard Statement GHS H Statement
H330-H227-H302
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P304+P340-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 202-862-5
RTECSNumber XT8650000
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only