missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7131281
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02245G 5 g
I022425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02245G Supplier TCI America Supplier No. I02245G
Only null left

Chemical Identifiers

CAS 622-78-6
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004819
InChI Key MDKCFLQDBWCQCV-UHFFFAOYSA-N
Synonym benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
PubChem CID 2346
ChEBI CHEBI:17484
IUPAC Name isothiocyanatomethylbenzene
SMILES C1=CC=C(C=C1)CN=C=S

Specifications

Boiling Point 243°C
Quantity 5 g
UN Number 3082
Formula Weight 149.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Isothiocyanate
EINECSNumber (3)-3243
RTECSNumber NX8250000
TSCA Yes