missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isothiocyanate, 98%

CAS: 622-78-6 | C8H7NS | 149.22 g/mol

$156.94 - $156.94

Chemical Identifiers

CAS 622-78-6
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.22
MDL Number MFCD00004819
InChI Key MDKCFLQDBWCQCV-UHFFFAOYSA-N
Synonym benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
PubChem CID 2346
ChEBI CHEBI:17484
IUPAC Name isothiocyanatomethylbenzene
SMILES C1=CC=C(C=C1)CN=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC402130250
SDS View Documents
Thermo Scientific Chemicals
402130250
25 g Glass bottle
Each for $156.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

622-78-6
149.22
MDKCFLQDBWCQCV-UHFFFAOYSA-N
2346
isothiocyanatomethylbenzene
C8H7NS
MFCD00004819
benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
CHEBI:17484
C1=CC=C(C=C1)CN=C=S

Specifications

622-78-6
100.0
1.1200g/mL
>110°C
97.5% min. (GC)
C8H7NS
CH2NCSC6H5
25 g
benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
C1=CC=C(C=C1)CN=C=S
149.22
CHEBI:17484
98%
Benzyl isothiocyanate, 98%
97.5
Yellow
243.0°C
Authentic
Glass bottle
1.6010 to 1.6040
MFCD00004819
1.12
MDKCFLQDBWCQCV-UHFFFAOYSA-N
isothiocyanatomethylbenzene
2346
149.22
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma sympto

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 210-753-9

RTECSNumber : NX8250000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only