missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isothiocyanate, 98%

Catalog No. AAA1500806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500806 5 g
AAA1500814 25 g
AAA1500822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500806 Supplier Thermo Scientific Chemicals Supplier No. A1500806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-78-6 | C8H7NS | 149.211 g/mol

Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 622-78-6
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004819
InChI Key MDKCFLQDBWCQCV-UHFFFAOYSA-N
Synonym benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
PubChem CID 2346
ChEBI CHEBI:17484
IUPAC Name isothiocyanatomethylbenzene
SMILES C1=CC=C(C=C1)CN=C=S

Specifications

Density 1.125
Boiling Point 242°C to 243°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.603
Quantity 5 g
UN Number UN2922
Beilstein 386135
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material Benzyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H351-H314-H318-H302-H312-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 210-753-9
RTECSNumber NX8250000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only